Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NVP BHG 712, Tocris Bioscience™

Catalog No. 44051
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-051 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-051 Supplier R&D Systems Supplier No. 4405/1
This item is not returnable. View return policy

Potent EphB4 kinase inhibitor

Selective inhibitor of EphB4 kinase; exhibits selectivity for EphB4 over more than 40 other kinases in vitro, including FGFR3. Inhibits EphB4 autophosphorylation in transiently transfected HEK 293 cells. Also inhibits VEGF-induced angiogenesis in vivo; thought to inhibit EphB4 forward signaling. Orally active.

Chemical Identifiers

CAS 940310-85-0
Molecular Formula C26H20F3N7O
Molecular Weight (g/mol) 503.49
MDL Number MFCD20272929
InChI Key ZCCPLJOKGAACRT-UHFFFAOYSA-N
Synonym nvp-bhg712, 4-methyl-3-1-methyl-6-pyridin-3-yl-1h-pyrazolo 3,4-d pyrimidin-4-ylamino-n-3-trifluoromethyl phenyl benzamide, 4-methyl-3-1-methyl-6-3-pyridinyl-1h-pyrazolo 3,4-d pyrimidin-4-yl amino-n-3-trifluoromethyl phenyl benzamide, 4-methyl-3-1-methyl-6-pyridin-3-yl pyrazolo 3,4-d pyrimidin-4-yl amino-n-3-trifluoromethyl phenyl benzamide, 4-methyl-3-1-methyl-6-pyridin-3-yl-1h-pyrazolo 3,4-d pyrimidin-4-yl amino-n-3-trifluoromethyl phenyl benzamide, nvpbhg712, nvp-bhg712/, nvp-bhg hplc, , nvp bhg712
PubChem CID 16747388
IUPAC Name 4-methyl-3-{[1-methyl-6-(pyridin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-yl]amino}-N-[3-(trifluoromethyl)phenyl]benzamide
SMILES CN1N=CC2=C(NC3=C(C)C=CC(=C3)C(=O)NC3=CC=CC(=C3)C(F)(F)F)N=C(N=C12)C1=CN=CC=C1

Specifications

Quantity 1 mg
Formula Weight 617.5
Percent Purity >98%
Chemical Name or Material NVP BHG 712
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.