Learn More
Selleck Chemical LLC NVP-CGM097 2mg 1313363-54-0 CGM-097
SDP
Supplier:  Selleck Chemical LLC S78752MG
Encompass_Preferred
NVP-CGM097 is a highly potent and selective MDM2 inhibitor with Ki value of 1.3 nM for hMDM2 in TR-FRET assay. It binds to the p53 binding-site of the Mdm2 protein, disrupting the interaction between both proteins, leading to an activation of the p53 pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5362
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.