Learn More
Selleck Chemical LLC PF-05175157
SDP
Supplier:  Selleck Chemical LLC S667225MG
Encompass_Preferred
PF-05175157 is a broad-spectrum effective ACC inhibitor with IC50 of 27 0 nM 33 0 nM 23 5 nM and 50 4 nM for human ACC1 human ACC2 rat ACC1 and rat ACC2 respectively
Catalog No. 50-000-01850
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.