Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NWD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561370 Supplier Invitrogen™ Supplier No. PA561370
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DQPWVFQCNP LEPDIFFVNH RKMSELLYHL TRCGKTDDLL YGIIMNFSWL YTMIKIGQFD KVLSDIELAY NYSQEKELKF LANTLRSIKN KVTAFP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NWD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NWD2
Gene Accession No. Q9ULI1
Gene Alias KIAA1239; Leucine-rich repeat and WD repeat-containing protein KIAA1239; NACHT and WD repeat domain containing 2; NACHT and WD repeat domain-containing protein 2; NWD2
Gene Symbols NWD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NWD2. Recombinant protein control fragment (Product #RP-100245).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57495
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.