Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569110 Supplier Invitrogen™ Supplier No. PA569110
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.

Could be involved in the export of mRNA from the nucleus to the cytoplasm. Could also have a role in polarized cytoplasmic transport and localization of mRNA in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXF5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NXF5
Gene Accession No. Q9H1B4
Gene Alias Nuclear RNA export factor 5; NXF5; TAPL1; TAPL-1; TAP-like protein 1
Gene Symbols NXF5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NXF5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55998
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.