Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570684 Supplier Invitrogen™ Supplier No. PA570684
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rat: 93%.

Retinitis pigmentosa (RP) is a disease that leads to blindness by degeneration of cone photoreceptors. Rods produce factors required for cone viability. The protein encoded by this gene is one of those factors and is similar to a truncated form of thioredoxin. This gene has been proposed to have therapeutic value against RP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXNL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nxnl1
Gene Accession No. Q96CM4
Gene Alias A930031O08Rik; nucleoredoxin like 1; nucleoredoxin-like 1; nucleoredoxin-like protein 1; NXNL1; RdCVF; rod-derived cone viability factor; thioredoxin-like 6; thioredoxin-like protein 6; Txnl6
Gene Symbols Nxnl1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human NXNL1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115861
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.