Learn More
Charles River Labs Cell Solutions NY-ESO-1 Peptide (157-165) (SLLMWITQC) 1mg, Cryopreserved
Supplier:  Charles River Labs Cell Solutions ANTIGEN1095
Encompass
This peptide is derived from the tumor associated antigen, NY-ESO-1, a commonly expressed tumor antigen of the cancer-testis family. The sequence for this peptide is SLLMWITQC from residues 157-165 and it binds to HLA-A 02:01.
Catalog No. 50-225-6094
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.