Learn More
Grainger POWER FIRST Cable Tie, 8 in Nominal Lg
Supplier:  Grainger 36J219
Encompass_Preferred
The Power First Cable Tie, 8" Nominal Length, Black, 100 Pieces is a cable tie made from nylon 6/6 with a tensile strength of 50 lbs. It has a self-locking feature and smooth, low profile head design. This cable tie has a temperature range of -40°F to 185°F and is UL recognized. It comes in a pack of 100 ties, each with a length of 8", width of 0.16", and thickness of 0.05". This product can be used for bundling wires and cables in various applications.
Catalog No. NC2712809
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.