Learn More
TARGETMOL CHEMICALS INC Nylidrin hydrochloride 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T017310MG
Encompass_Preferred
Nylidrin hydrochloride was an effective inhibitor of the IgG1-mediated release of histamine from passively sensitized guinea pig lung slices, and of the IgE-mediated release of histamine from passively sensitized rat peritoneal mast cells and human basophils.
Catalog No. NC1914769
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.