Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Nylon 66 Filter Membranes

Catalog No. 111021712 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-1712 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1712 Supplier MilliporeSigma™ Supelco™ Supplier No. 58067
Only null left
Add to Cart
Add to Cart

Protect the instrument and columns by removing particles and gases from solvents and other mobile phase components.

  • Compatible with all solvents commonly used in HPLC. For use with sink aspirator.
  • Used in filtering the reagents such as acetonitrile and methanol used as mobile phase in the determination of aflatoxins in rice samples using high-performance liquid chromatography with fluorescence detection (HPLC-FL)
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 47 mm
Pore Size 0.45 μm
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.