Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Nylon Biopsy Bags
GSA_VA

Catalog No. 6774011 Shop All Epredia Products
Change view
Click to view available options
Size:
Small
Medium
Large
Quantity:
1000/Pk.
200/Pk.
500/Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Quantity
6774011 Large 500/Pk.
6774009 Medium 1000/Pk.
6774010 Small 1000/Pk.
6774012 Medium 200/Pk.
6774013 Medium 500/Pk.
6774014 Small 200/Pk.
6774015 Small 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 6774011 Supplier Epredia™ Supplier No. 6774011
Add to Cart
Add to Cart

Ensure unrestricted fluid movement around tissue with Epredia™ Nylon Biopsy Bags.

  • Reduce risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag

Dimensions:

  • Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Biopsy Bag
Material Nylon
Color Blue
Length (Metric) 75 mm
Width (Metric) 85 mm
Length (English) 3 in.
Width (English) 3.5 in.
Size Large
Quantity 500/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.