Learn More
Warner Instruments™ Nylon Mesh Kit
Catalog No. BTX640198 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
BTX640198 10/Pk.
1 options
Catalog No. BTX640198 Supplier Warner Instruments™ Supplier No. 640198
Very fine 7 x 7 cm nylon mesh supports and holds slices in recording and imaging chambers and comes in a pack of 10.
  • The NYL/MESH nylon mesh kit can be used in conjunction with Warner Instruments slice anchors or other wire or plastic holders.
  • These meshes can be used to either support tissue slices or to hold slices in place in saline solutions. This could be helpful with many recording and imaging techniques. Each kit includes sections of nylon mesh in two sizes. Meshes are 0.3 mm and 1.13 mm mesh opening and are 0.2 and 0.12 mm thick respectively. Five 7 x 7 cm squares of each size are supplied.
  • These meshes can be used to either support tissue slices or to hold slices in place in saline solutions. This could be helpful with many recording and imaging techniques.
  • Each kit includes sections of nylon mesh in two sizes. Meshes are 0.3 mm and 1.13 mm mesh opening and are 0.2 and 0.12 mm thick respectively. Five 7 x 7 cm squares of each size are supplied.
Specifications
Quantity 10/Pk.