Learn More
Braintree Scientific Nylon Monofilament, Non-absorbable Suture, 7-0 100 yds
SDP
Supplier:  Braintree Scientific SUTM103
Encompass
Monofilament Nylon Suture Nylon Monofilament, Non-absorbable Suture. Deknatel's reliable nylon suture is known for its uniform diameter, flexibility, and outstanding knot security. Material: Nylon type 6,6. Coating: None. Characteristics: Nylon exhibits uniform diameter of suture, minimal memory, and outstanding and consistent knot security. Please note: Nylon suture should not be used where permanent retention of tensile strength is required. Indications: Nylon suture is indicated for use in general soft tissue approximation and/or ligation, including use in cardiovascular, neurological, and ophthalmic procedures. Sizes: USP 2 through 8-0. Construction: Monofilament. Configuration: Non-sterile spool. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5978
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.