Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ NZ Bovine Transferrin (Holo), Heat Treated

Catalog No. 0215233591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-33591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-152-33591 Supplier MP Biomedicals™ Supplier No. 0215233591
Add to Cart
Add to Cart

Iron-rich product supplied as a heat-treated, lyophilized powder. Purified from New Zealand-sourced bovine plasma, ISO quality assures complete traceability and consistent high quality.

  • Chromatographically purified from New Zealand-sourced bovine plasma
  • An ISO quality system assures complete traceability and consistent high quality
  • Transferrin (HOLO) is an iron-rich product supplied as a heat-treated, lyophilised powder
TRUSTED_SUSTAINABILITY

Specifications

Concentration Total Protein (anhydrous) ≥95% w/w total protein
Formulation Lyophilized
Quantity 1 g
Regulatory Status RUO
Species Bovine
Classification ISO
Protein Family Transferrin
Form Powder
Purity or Quality Grade Cell Culture Grade
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.