Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-1918, R&D Systems™
SDP

Catalog No. 228810 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-881-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-881-0 Supplier R&D Systems Supplier No. 2288/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Silent antagonist for putative abnormal-CBD (Cat. No. 1297) receptor

Selective, silent antagonist of a putative endothelial anandamide receptor distinct from CB1 or CB2 receptors. Inhibits vasodilation and cell migration induced by abnormal-cannabidiol (abn-CBD; Cat. No. 1297).

Chemical Identifiers

CAS 536697-79-7
Molecular Formula C19H26O2
Molecular Weight (g/mol) 286.415
InChI Key ICHJMVMWPKLUKT-JKSUJKDBSA-N
Synonym 1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl benzene, 1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl-benzene, 1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-prop-1-en-2-yl cyclohex-2-en-1-yl benzene, benzene,1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl
PubChem CID 40469923
IUPAC Name 1,3-dimethoxy-5-methyl-2-[(1R,6R)-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl]benzene
SMILES CC1=CC(C(CC1)C(=C)C)C2=C(C=C(C=C2OC)C)OC

Specifications

Quantity 10 mg
Formula Weight 286.41
Percent Purity >97%
Product Line Tocris
Chemical Name or Material O-1918
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.