Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime 98.0+%, TCI America™
SDP

Catalog No. P1190100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1190100MG 100 mg
P11901G 1 g
P11905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P1190100MG Supplier TCI America Supplier No. P1190100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86356-73-2
Molecular Formula C8H4F5NO
Molecular Weight (g/mol) 225.118
MDL Number MFCD00191477
InChI Key SRTQFRQWTUMMTC-UHFFFAOYSA-N
PubChem CID 602486
IUPAC Name N-[(2,3,4,5,6-pentafluorophenyl)methoxy]methanimine
SMILES C=NOCC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color White
Quantity 100 mg
Formula Weight 225.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.