Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-[2-Oxo-1(2H)-pyridyl]-N,N,N',N'-tetramethyluronium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. O03901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03901G 1 g
O03905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03901G Supplier TCI America Supplier No. O03901G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 125700-71-2
Molecular Formula C10H16BF4N3O2
Molecular Weight (g/mol) 297.06
MDL Number MFCD00075475
InChI Key CZQGINAUZYECAI-UHFFFAOYSA-N
Synonym tptu, 2-2-pyridon-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, o-1,2-dihydro-2-oxo-1-pyridyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, tptu o-1,2-dihydro-2-oxo-pyridyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 1,1,3,3-tetramethyl-2-2-oxopyridin-1 2h-yl uronium tetrafluoroborate, o-2-oxo-1 2h pyridyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, dimethylamino 2-oxopyridin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, pubchem12779, o-1,2-dihydro-2-oxo-1-pyridyl-n,n,n',n'-tetramethyluronium tetrafluoroborate tptu, acmc-1c5pe
PubChem CID 2733204
IUPAC Name [(dimethylamino)[(2-oxo-1,2-dihydropyridin-1-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C(ON1C=CC=CC1=O)=[N+](C)C

Specifications

Melting Point 137°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 297.06
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-[2-Oxo-1(2H)-pyridyl]-N,N,N′,N′-tetramethyluronium Tetrafluoroborate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.