Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. D326325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D326325G 25 g
D32635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D326325G Supplier TCI America Supplier No. D326325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 125700-69-8
Molecular Formula C12H16BF4N5O2
Molecular Weight (g/mol) 349.10
MDL Number MFCD00077730
InChI Key FOBCPCIJLQTYBT-UHFFFAOYSA-N
Synonym tdbtu, n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 1,1,3,3-tetramethyl-2-4-oxobenzo d 1,2,3 triazin-3 4h-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, tdbtu; n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, dimethylamino 4-oxo-1,2,3-benzotriazin-3-yl oxy methylidene dimethylazanium tetrafluoroborate, pubchem12760, acmc-1cac0, d04fkx
PubChem CID 10882602
IUPAC Name [(dimethylamino)[(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C2C1=O)=[N+](C)C

Specifications

Color White
Quantity 25 g
UN Number 1759
Formula Weight 349.10
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N′,N′-tetramethyluronium Tetrafluorobo
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.