Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. C192625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C192625G 25 g
C19265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C192625G Supplier TCI America Supplier No. C192625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 330641-16-2
Molecular Formula C11H15BClF4N5O
Molecular Weight (g/mol) 355.53
MDL Number MFCD04973270
InChI Key GBGVQFJZGHBZMC-UHFFFAOYSA-N
Synonym tctu, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-6-chloro-1h-benzotriazol-1-yl uronium tetrafluoroborate, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborat e, 6-chlorobenzotriazol-1-yl oxy-dimethylamino methylene-dimethyl-ammonium tetrafluoroborate, tctu; 1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium tetrafluoroborate, 6-chloro-1,2,3-benzotriazol-1-yl oxy dimethylamino methylidene dimethylazanium tetrafluoroborate, 1h-benzotriazolium, 1-bis dimethylamino methylene-5-chloro-, 3-oxide tetrafluoroborate 1-1:1, acmc-1agto, ksc569q6h
PubChem CID 24898642
IUPAC Name tetrafluoroboranuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C

Specifications

Melting Point 204°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 355.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(6-Chlorobenzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.