Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Acetotoluidine 98.0+%, TCI America™
SDP

Catalog No. A0063500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0063500G 500 g
A006325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0063500G Supplier TCI America Supplier No. A0063500G

Chemical Identifiers

CAS 120-66-1
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00014961
InChI Key BPEXTIMJLDWDTL-UHFFFAOYSA-N
Synonym o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide
PubChem CID 8443
IUPAC Name N-(2-methylphenyl)acetamide
SMILES CC1=CC=CC=C1NC(=O)C

Specifications

Melting Point 110°C
Color Gray-White
Boiling Point 296°C
Quantity 500 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material o-Acetotoluidine
RTECSNumber AN2900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.