Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-O-Acetyl-D-mandelic Acid 98.0+%, TCI America™
SDP

Catalog No. A145325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A145325G 25 g
A14535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A145325G Supplier TCI America Supplier No. A145325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51019-43-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004249
InChI Key OBCUSTCTKLTMBX-UHFFFAOYNA-N
Synonym r-2-acetoxy-2-phenylacetic acid, r---alpha-acetoxyphenylacetic acid, --o-acetyl-d-mandelic acid, r---o-acetylmandelic acid, r-alpha-acetoxyphenylacetic acid, 2r-2-acetyloxy-2-phenylacetic acid, benzeneacetic acid, alpha-acetyloxy-, alphar, r-acetyloxy phenyl acetic acid, s-+-o-acetyl-l-mandelic acid, r---a-acetylmandelic acid
PubChem CID 2733814
IUPAC Name 2-(acetyloxy)-2-phenylacetic acid
SMILES CC(=O)OC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 98°C
Color White
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-O-Acetyl-D-mandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.