Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Acetyl-N-carbobenzoxyhydroxylamine 98.0+%, TCI America™
SDP

Catalog No. A21931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21931G 1 g
A21935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21931G Supplier TCI America Supplier No. A21931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 180798-01-0
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
InChI Key YEIRDDKOGLZHLP-UHFFFAOYSA-N
Synonym O-Acetyl-N-Cbz-hydroxylamine, N-Acetoxycarbamic Acid Benzyl Ester, Benzyl N-Acetoxycarbamate
PubChem CID 10954788
IUPAC Name phenylmethoxycarbonylamino acetate
SMILES CC(=O)ONC(=O)OCC1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 209.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Acetyl-N-carbobenzoxyhydroxylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.