Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Anisidine, 99%

Catalog No. AAA102880B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA102880B 1000 g
AAA1028830 250 g
AAA102880I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA102880B Supplier Thermo Scientific Chemicals Supplier No. A102880B
Only null left
Add to Cart

CAS: 90-04-0 | C7H9NO | 123.155 g/mol

o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007688
InChI Key VMPITZXILSNTON-UHFFFAOYSA-N
Synonym o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
PubChem CID 7000
ChEBI CHEBI:82288
IUPAC Name 2-methoxyaniline
SMILES COC1=CC=CC=C1N

Specifications

Melting Point 5°C to 6°C
Density 1.092
Boiling Point 224°C to 225°C
Flash Point 98°C (208°F)
Assay Percent Range 99%
Refractive Index 1.574
Quantity 1000 g
UN Number UN2431
Beilstein 386210
Sensitivity Light sensitive
Merck Index 14,667
Solubility Information Soluble in dilute inorganic acid,alcohol,diethyl ether. Not miscible in water.
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material o-Anisidine
Show More Show Less
Hazard Category H301+H311-H341-H350-H373
Hazard Statement GHS H Statement
H301-H311-H331-H350-H341
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P310-P302+P352-P308+P313-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANISIDINES
EINECSNumber 201-963-1
RTECSNumber BZ5410000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.