Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzyl-DL-serine 98.0+%, TCI America™
SDP

Catalog No. B086010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B086010G 10 g
B08601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B086010G Supplier TCI America Supplier No. B086010G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5445-44-3
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00021724,MFCD00065936
InChI Key IDGQXGPQOGUGIX-UHFFFAOYNA-N
Synonym o-benzyl-dl-serine, 2-amino-3-benzyloxy propanoic acid, o-benzylserine, h-dl-ser bzl-oh, 3-benzyloxy-dl-alanine, benzylserine, 2-amino-3-phenylmethoxy propanoic acid, o-benzyl-serine, acmc-1artk, spectrum2_001351
PubChem CID 4573
IUPAC Name 2-amino-3-(benzyloxy)propanoic acid
SMILES NC(COCC1=CC=CC=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 195.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzyl-DL-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.