Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ O-Cedar™ Power Tip Bottle Brush

Catalog No. 19200746 Shop All Vileda Professional Products
Change view
Click to view available options
O-Cedar™ Power Tip Bottle Brush
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-200-746 O-Cedar™ Power Tip Bottle Brush
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-200-746 Supplier Vileda Professional™ Supplier No. 148384
This item is not returnable. View return policy

Extra-long handle and 360° bristles reach deep into tall glasses and bottles

  • Strong bristles at tip to provide non-scratch scrubbing power
  • Ergnomic, sturdy handle is rubberized for comfort and better grip when wet
  • Specially designed thumb spot for additional pressure on tough spots
  • Designed to keep hands dry and clean
  • Bristles have excellent bend recovery and resist stains

Specifications

Product Type Bottle Brush
Dimensions (L x W x H) 14 x 2 x 2 in.
For Use With (Application) Bottles
Length (English) 14 in.
Material Nylon
Quantity 12/cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.