Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresolphthalein Complex, Solstice

Catalog No. 60047257 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-257 1 g Glass Bottle
60-047-258 25 g Glass Bottle
60-047-259 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-257 Supplier Solstice Supplier No. 640001G

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2411-89-4
Molecular Formula C32H32N2O12
Molecular Weight (g/mol) 636.61
MDL Number MFCD00005911
InChI Key IYZPEGVSBUNMBE-UHFFFAOYSA-N
PubChem CID 75485
IUPAC Name 2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Melting Point 179°C to 182°C
Boiling Point >260°C
UN Number NONH for all modes of transport
Beilstein 381994
Formula Weight 636.6g/mol
Odor Unpleasant
Packaging Glass Bottle
Color Yellow
Quantity 1 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein Complexone
Show More Show Less
RTECSNumber 219-318-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.