Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam o-Cresolphthalein Complexon, Alkaline Earth Metal Indicator, 25 g

Catalog No. 502704766
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4766 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4766 Supplier Abcam Supplier No. AB14627525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alkaline earth metal indicator used for colorimetric estimation of calcium in serum.

  • Molecular Weight: 636.6 g/mol
  • Application: Alkaline earth metal indicator; used for colorimetric estimation of calcium in serum
  • Composition: 100% Phthalein purple

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Calcium
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate Calcium
Format Bottle
Buffer 100% phthalein purple
Content And Storage Store at ambient temperature.
Form Powder
Detection Method Colorimetric
Formulation 100% Phthalein purple
Sample Type Serum
Quantity 25 g
For Use With (Application) Colorimetric estimation of calcium in serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.