Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresolphthalein, Indicator Grade, 98%, Thermo Scientific™

Catalog No. AC110601000
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC110601000 100 g
AC110600100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110601000 Supplier Thermo Scientific Chemicals Supplier No. 110601000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-27-0
Molecular Formula C22H18O4
Molecular Weight (g/mol) 346.38
MDL Number MFCD00005912
InChI Key CPBJMKMKNCRKQB-UHFFFAOYSA-N
PubChem CID 68995
IUPAC Name 3,3-bis(4-hydroxy-3-methylphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES CC1=CC(=CC=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Beilstein 18, 153
Merck Index 15, 2565
Infrared Spectrum Authentic
Formula Weight 346.38
Loss on Drying 1% max.
Percent Purity ≥97.5% (UV)
Grade Indicator
Visual Transition Interval from pH 8.0 (colorless) to pH 9.8 (red)
Color Beige to White or Tan
Quantity 100 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein, 98%
Show More Show Less
EINECSNumber 209-881-8
RTECSNumber SM8390000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.