accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOHOH3C

CAS RN 596-27-0

IUPAC Name: 3,3-bis(4-hydroxy-3-methylphenyl)-1,3-dihydro-2-benzofuran-1-one
Synonyms: cresolphthalein
Molecular Weight (g/mol): 346.38
Molecular Formula: C22H18O4
InChi Key: CPBJMKMKNCRKQB-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (6)
  • (1)
Physical Form 
  • (1)
  • (5)
  • (1)
Color 
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)