Learn More
Strem, An Ascensus Company CAS 1289463-93-9. 100mg. Bis(2-trifluoromethylphenyl)(3-sulfonatophenyl)phosphine sodium salt min. 97% o-DAN2PHOS. MFCD27978401
SDP
Supplier:  Strem, An Ascensus Company 150579100MG
Encompass_Preferred
CAS 1289463-93-9. 100mg. Bis(2-trifluoromethylphenyl)(3-sulfonatophenyl)phosphine sodium salt min. 97% o-DAN2PHOS. MFCD27978401. Molecular Weight 500.33. Molecular Formula C20H12F6NaO3PS. Color/form white pwdr. Strem 15-0579. http//www.strem.com/catalog/v/15-0579/
Catalog No. 50-211-0739
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.