Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (18B10.C7)
GREENER_CHOICE

Catalog No. PIMA1038 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1038 Supplier Invitrogen™ Supplier No. MA1038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-038 has been successfully used in Western blot and immunoprecipitation applications with human samples. In Western blot, MA1-038 detects proteins with O-GlcNAc post-translational modification, and therefore will detect multiple specific bands.

O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen O-linked N-acetylglucosamine (O-GlcNAc)
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 18B10.C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene OGT
Gene Accession No. O15294
Gene Alias GlcNAc; HINCUT-1; HRNT1; O-GLCNAC; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; OGT; O-linked N-acetylglucosamine (GlcNAc) transferase; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase
Gene Symbols OGT
Host Species Mouse
Immunogen SANM-conjugated linear peptide corresponding to human O-GlcNAc.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8473
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.