Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O,O'-Bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol

Catalog No. 14529100GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-529-100GF 100 g
14-529-500GF 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-529-100GF Supplier Sigma Aldrich Supplier No. 14529100GF
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(O,O' -bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol (Jeffamine(R) ED-2003) is a functional oligomer with a high content of polypropylene glycol (PEO). {3}Jeffamine(R)-ßED-2003 may be used for the solubilisation of CdSe-based quantum dots (QDs). Functionalization of QDs makes it easier for them to bind with proteins. {2} Jeffamine(R)-ßED-2003 may be used for the insitu fabrication of PEO/silica membranes.{3}Jeffamine is a registered trademark of Huntsman Petrochemical, LLC

Specifications

CAS 65605-36-9
Linear Formula CH3CH(NH2)CH2[OCH(CH3)CH2]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
MDL Number MFCD00147794
Quantity 100 g
Synonym Jeffamine™ ED-2003
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.