Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ o-Phenanthroline Monohydrate
SCI_ED

Catalog No. s25461c Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Reagent Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25461C 5 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25461C
Only null left
Add to Cart
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PubChem CID 21226
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Packaging Reagent Grade
Linear Formula C12H8N2· H2O
Quantity 5 g
Formula Weight 198.21
Grade Reagent
Chemical Name or Material 1, 10-Phenanthroline Monohydrate
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.