Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phenylenediamine, 98%

Catalog No. p-3737193
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130552500 250 g Glass bottle
AC130550010 1 kg Glass bottle
AC130550050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130552500 Supplier Thermo Scientific Chemicals Supplier No. 130552500
Only null left
Add to Cart

CAS: 95-54-5 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-54-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007721
InChI Key GEYOCULIXLDCMW-UHFFFAOYSA-N
Synonym o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
PubChem CID 7243
ChEBI CHEBI:34043
IUPAC Name benzene-1,2-diamine
SMILES C1=CC=C(C(=C1)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 100.0°C to 103.0°C
Color White to Beige
Boiling Point 256.0°C to 258.0°C
Flash Point 136°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(NH2)2
Quantity 250 g
Beilstein 13, IV, 38
Fieser 01,834; 08,393
Merck Index 15, 7396
Solubility Information Solubility in water: 54g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 108.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material o-Phenylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause an allergic skin reaction.
Suspected of causing genetic
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a posit
EINECSNumber 202-430-6
Recommended Storage This product tends to darken during storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.