Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phospho-L-Threonine, 99.6%, MP Biomedicals™

Catalog No. ICN15624283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15624283 250 mg
ICN15624290 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15624283 Supplier MP Biomedicals, Inc Supplier No. 0215624283

Chemical Identifiers

CAS 1114-81-4
Molecular Formula C4H10NO6P
Molecular Weight (g/mol) 199.10
MDL Number MFCD00069578,MFCD00135662
InChI Key USRGIUJOYOXOQJ-UHFFFAOYNA-N
Synonym phosphothreonine, o-phospho-l-threonine, o-phosphothreonine, l-threonine phosphate, threoninium dihydrogen phosphate, l-threonine o-phosphate, h-thr po3h2-oh, l-threonine o-3-phosphate, o3-phosphothreonine, 2s,3r-2-amino-3-phosphonooxy butanoic acid
PubChem CID 3246323
ChEBI CHEBI:37525
IUPAC Name 2-amino-3-(phosphonooxy)butanoic acid
SMILES CC(OP(O)(O)=O)C(N)C(O)=O

Specifications

Color White
Assay Percent Range 0.996
Quantity 250 mg
Formula Weight 199.1
Physical Form Powder
Chemical Name or Material o-Phospho-L-Threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.