Learn More
AdipoGen o-Phthaldialdehyde
SDP
Supplier:  AdipoGen CDXO0008G005
Encompass_Preferred
Chemical. CAS 643-79-8. Formula C8H6O2. MW 134.13. Synthetic Flow cytometry measurement tool. Fluorescence probe for glutathione and glutathione disulfide levels, primary amines, peptides and biological thiols. Reacts with primary amines to produce a product that emits a highly fluorescent blue colour. Spectral data lambdaex 334 nm lambdaem 455 nm Thiol Adduct. Used for the quick visualization of histamine, characterized by the appearance of a yellow stain. Used for precolumn derivatization of amino acids for HPLC separation. High-level disinfectant/sterilant for endoscopes, thermometers, rubber and plastic equipment, and medical/surgical instruments that cannot be sterilized by heat, based on its antimicrobial efficacy and low inhalation toxicity.
Catalog No. 50-256-0350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.