Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ O.R. Grade Mayo Dissecting Scissors - Serrated Blade, Anti-glare

Catalog No. 21124946 Shop All Surgical Design Products
Change view
Click to view available options
Includes:
Curved, Serrated Blade
Straight, Serrated Blade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-124-946 Curved, Serrated Blade
21-124-944 Straight, Serrated Blade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-124-946 Supplier Surgical Design™ Supplier No. ORG124SSR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O.R.-grade Mayo scissors with one serrated blade support controlled cutting of dense tissue.

  • Length: 6.75 in.
  • Configuration Options: Straight or Curved
  • One serrated blade provides superior bite and prevents tissue slippage
  • Heavy-duty construction ideal for firm dissection and thick tissue cutting
  • Precision-ground edges ensure smooth, consistent performance
  • Autoclavable stainless steel suitable for repeated sterilization
  • Ideal for surgery, clinical use, laboratory settings, and autopsy/postmortem applications

Specifications

Product Type Dissecting Scissors
Material Stainless Steel
Includes Curved, Serrated Blade
Length (English) 6.75 in.
Length (Metric) 16.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.