Learn More
Cytiva O-Ring-117 Viton Black
Supplier: Cytiva 83475
Large Filtration Area for Easy Operation and Fast Liquid Flow-Economical for small-volume (10 to 100 mL) filtration.-Accepts 25 mm filter discs.-Luer inlet and outlet fittings provide easy connections.-Delrin (acetal resin) construction provides broad chemical compatibility and material strength. This product is not approved for use in medical, clinical, surgical or other patient applications.