Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ O-ring
GREENER_CHOICESDP

Catalog No. 31502296 Shop All Kemtech Products
Change view
Click to view available options
Diameter (Metric):
3.7 mm
4.5 mm
4.72 mm
5.23 mm
6 mm
6.07 mm
7.65 mm
9.2 mm
10.8 mm
12.30 mm
12.4 mm
18.7 mm
21.8 mm
26.6 mm
29.7 mm
40.9 mm
50.4 mm
56.5 mm
59.9 mm
81.9 mm
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
31-502-296 29.7 mm
31-501-937 3.7 mm
31-502-293 4.5 mm
31-501-950 4.72 mm
31-501-941 5.23 mm
31-501-942 6 mm
31-501-938 6.07 mm
31-502-294 7.65 mm
31-501-943 9.2 mm
31-501-939 10.8 mm
31-501-940 12.30 mm
31-501-944 12.4 mm
31-501-945 18.7 mm
31-501-947 21.8 mm
31-501-946 26.6 mm
31-501-949 40.9 mm
31-501-951 50.4 mm
31-501-953 56.5 mm
31-501-952 59.9 mm
31-502-297 81.9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 31-502-296 Supplier Synthware™ Supplier No. R217
Add to Cart
Add to Cart

For use with solvent and chemical compatibility.

  • O-rings for solvent and chemical compatibility

Specifications

Color Beige
For Use With (Equipment) #25 O-Ring Joint
Material Viton
Type O-Ring
Diameter (English) 1.17 in.
Diameter (Metric) 29.7 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.