Learn More
Beckman Coulter GS-6 Centrifuge, Technical Bulletin, TB-003
Supplier:  Beckman Coulter 345366
Encompass
The Beckman Coulter GS-6R is a refrigerated benchtop centrifuge with a max speed of 14,000 rpm and RCF of 21,382 x g. It holds 6 x 85mL tubes and offers quiet operation with a brushless motor. It features programmable acceleration/deceleration, a vacuum fluorescent display, rotor identification, and maintains 4-25°C. Safety includes an imbalance detector, lid interlock, and overspeed detector, suitable for various research applications.
Catalog No. NC1244362
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.