Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ O-ring Kits for High Vacuum Valve Plugs
GREENER_CHOICESDP

Catalog No. 31502056 Shop All Kemtech Products
Change view
Click to view available options
For Use With (Equipment):
S540004
S540008
S540012
Includes:
R007(x1) and R010(x2)
R108(x1) and R110(x2)
R110(x1) and R112(x2)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes
31-502-056 S540004 R007(x1) and R010(x2)
31-502-057 S540008 R108(x1) and R110(x2)
31-502-058 S540012 R110(x1) and R112(x2)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-056 Supplier Synthware™ Supplier No. R7010
Add to Cart
Add to Cart

For use with solvent and chemical compatibility.

Specifications

Color Brown
For Use With (Equipment) S540004
Material Viton
Includes R007(x1) and R010(x2)
Quantity 3 Pk.
Type O-Ring Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.