Learn More
Vernier™ O2 Gas Sensor to Spirometer Adapter
SDPSCI_ED
Catalog No. S16227ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
CO2 Gas Sensor, O2 Gas Sensor, Spirometer
Catalog No. For Use With (Equipment)
S16227ND CO2 Gas Sensor, O2 Gas Sensor, Spirometer
1 options
Catalog No. S16227ND Supplier Vernier™ Supplier No. O2SPR
Add to Cart
Add to Cart
Includes
O2 Gas Sensor to Spirometer Adapter
This adapter facilitates the connection between an Oxygen Gas Sensor and a Spirometer when measuring the oxygen gas concentration and flow rate of exhaled air. It is used in .
This is the power of electromagnetic radiation in the spectral range that is used by plants for photosynthesis (400–700 nm). It features a waterproof sensor head and can be used to measure PPFD from sunlight and electric light sources. This sensor is ideal for experiments investigating photosynthesis and primary productivity and can be used in many agricultural and environmental science applications.
Warranty and Services
This is a consumable and no warranty is included.
Specifications
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) CO2 Gas Sensor, O2 Gas Sensor, Spirometer