Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-tert-Butyl-L-threonine tert-Butyl Ester 96.0+%, TCI America™
SDP

Catalog No. B334025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B334025G 25 g
B33405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B334025G Supplier TCI America Supplier No. B334025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5854-78-4
Molecular Formula C12H25NO3
Molecular Weight (g/mol) 231.34
MDL Number MFCD00077109
InChI Key PPDIUNOGUIAFLV-UHFFFAOYNA-N
Synonym H-Thr(tBu)-OtBu
PubChem CID 7018832
IUPAC Name tert-butyl 2-amino-3-(tert-butoxy)butanoate
SMILES CC(OC(C)(C)C)C(N)C(=O)OC(C)(C)C

Specifications

Boiling Point 70°C
Quantity 25 g
Formula Weight 231.34
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material O-tert-Butyl-L-threonine tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.