Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(Tetrahydropyran-2-yl)hydroxylamine 98.0+%, TCI America™
SDP

Catalog No. T25981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25981G 1 g
T25985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25981G Supplier TCI America Supplier No. T25981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6723-30-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD01321374
InChI Key NLXXVSKHVGDQAT-UHFFFAOYNA-N
Synonym o-tetrahydro-2h-pyran-2-yl hydroxylamine, o-tetrahydropyran-2-yl-hydroxylamine, o-oxan-2-yl hydroxylamine, o-tetrahydropyran-2-yl hydroxylamine, 2-aminooxy tetrahydro-2h-pyran, o-tetrahydro-pyran-2-yl-hydroxylamine, o-tetrahydropyran-2-ylhydroxylamine, o-tertrahydroxy-pyran-2-yl-hydroxylamine, hydroxylamine, o-tetrahydro-2h-pyran-2-yl, h2nothp
PubChem CID 5142091
IUPAC Name O-(oxan-2-yl)hydroxylamine
SMILES NOC1CCCCO1

Specifications

Melting Point 36°C
Color White
Boiling Point 81°C
Quantity 1 g
Formula Weight 117.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material O-(Tetrahydropyran-2-yl)hydroxylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.