Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Toluic Acid 98.0+%, TCI America™
SDP

Catalog No. T0292100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0292100G 100 g
T029225G 25 g
T0292500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0292100G Supplier TCI America Supplier No. T0292100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 118-90-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002477
InChI Key ZWLPBLYKEWSWPD-UHFFFAOYSA-N
Synonym o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
PubChem CID 8373
ChEBI CHEBI:36632
IUPAC Name 2-methylbenzoic acid
SMILES CC1=CC=CC=C1C(=O)O

Specifications

Melting Point 104°C
Color White-Yellow
Boiling Point 259°C
Quantity 100 g
Formula Weight 136.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Toluic Acid
EINECSNumber (3)-1285
RTECSNumber XU1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.