Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolunitrile 98.0+%, TCI America™
SDP

Catalog No. T030625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T030625G 25 g
T0306500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T030625G Supplier TCI America Supplier No. T030625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 529-19-1
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00001792
InChI Key NWPNXBQSRGKSJB-UHFFFAOYSA-N
Synonym o-tolunitrile, 2-cyanotoluene, o-toluonitrile, benzonitrile, 2-methyl, o-cyanotoluene, o-tolylnitrile, o-toluic nitrile, 2-methylbenzenecarbonitrile, o-methylbenzonitrile, 2-tolunitrile
PubChem CID 10721
IUPAC Name 2-methylbenzonitrile
SMILES CC1=CC=CC=C1C#N

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Tolunitrile
EINECSNumber (3)-1804
RTECSNumber XV0600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.