Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Toluoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T0310100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0310100G 100 g
T031025G 25 g
T0310500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0310100G Supplier TCI America Supplier No. T0310100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 933-88-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000668
InChI Key GPZXFICWCMCQPF-UHFFFAOYSA-N
Synonym o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
PubChem CID 70276
IUPAC Name 2-methylbenzoyl chloride
SMILES CC1=CC=CC=C1C(Cl)=O

Specifications

Color Yellow
Boiling Point 213°C
Quantity 100 g
UN Number 3265
Formula Weight 154.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material o-Toluoyl Chloride
EINECSNumber (3)-1399&(3)-2972
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.