Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. T0881250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0881250G 250 g
T088125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0881250G Supplier TCI America Supplier No. T0881250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 644-36-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00004328
InChI Key RZWGTXHSYZGXKF-UHFFFAOYSA-N
Synonym o-tolylacetic acid, 2-methylphenylacetic acid, 2-methylphenyl acetic acid, 2-2-methylphenyl acetic acid, 2-o-tolyl acetic acid, 2-tolylacetic acid, 2-o-tolylacetic acid, o-methylphenylacetic acid, o-tolyacetic acid
PubChem CID 69519
IUPAC Name 2-(2-methylphenyl)acetic acid
SMILES CC1=CC=CC=C1CC(=O)O

Specifications

Melting Point 89°C
Color White
Quantity 250 g
Formula Weight 150.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Tolylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.