Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylylene Dicyanide 98.0+%, TCI America™
SDP

Catalog No. X005025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X005025G 25 g
X00505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X005025G Supplier TCI America Supplier No. X005025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 613-73-0
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00001905
InChI Key FWPFXBANOKKNBR-UHFFFAOYSA-N
Synonym 1,2-phenylenediacetonitrile, o-phenylenediacetonitrile, 1,2-bis cyanomethyl benzene, o-xylylene dicyanide, 2-2-cyanomethyl phenyl acetonitrile, o-benzenediacetonitrile, 2,2'-1,2-phenylene diacetonitrile, o-bis cyanomethyl benzene, 1,2-benzenediacetonitrile, o-benzendiacetonitrile
PubChem CID 69180
IUPAC Name 2-[2-(cyanomethyl)phenyl]acetonitrile
SMILES N#CCC1=CC=CC=C1CC#N

Specifications

Melting Point 59°C
Quantity 25 g
UN Number 3439
Formula Weight 156.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material o-Xylylene Dicyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.