Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylylene Oxide 97.0+%, TCI America™
SDP

Catalog No. X005325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X005325G 25 g
X00535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X005325G Supplier TCI America Supplier No. X005325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 496-14-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005932
InChI Key SFLGSKRGOWRGBR-UHFFFAOYSA-N
Synonym phthalan, 1,3-dihydroisobenzofuran, isocoumaran, isobenzofuran, 1,3-dihydro, o-xylylene oxide, 2-oxaindan, 1,3-dihydro isobenzofuran, unii-2r6ntq7y6g, ccris 1557, 2r6ntq7y6g
PubChem CID 10327
IUPAC Name 1,3-dihydro-2-benzofuran
SMILES C1C2=CC=CC=C2CO1

Specifications

Color Colorless
Boiling Point 192°C
Quantity 25 g
Formula Weight 120.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Xylylene Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.